Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIH-1/HIF-1AN Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB100428H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100428H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100428H Supplier Novus Biologicals Supplier No. NB100428H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FIH-1/HIF-1AN Polyclonal specifically detects FIH-1/HIF-1AN in Human, Mouse, Rat samples. It is validated for Electron Microscopy, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen FIH-1/HIF-1AN
Applications Electron Microscopy, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated
Formulation PBS with No Preservative
Gene Accession No. Q9NWT6
Gene Alias DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase
Gene Symbols HIF1AN
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Factor Inhibiting HIF-1 (between residues 300 and the C-terminus). [UniProt# Q9NWT6]
Molecular Weight of Antigen 40.3 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55662
Test Specificity Human, rat and mouse (PMID 19720742).
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.