Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filaggrin Antibody (FLG/1563), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB009109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009109 Supplier Novus Biologicals Supplier No. NBP275774IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Filaggrin Monoclonal specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Filaggrin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FLG/1563
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Gene Symbols FLG
Host Species Mouse
Immunogen Recombinant human Filaggrin protein fragment (around aa 998-1104) (exact sequence is proprietary) (Uniprot: P20930)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Test Specificity Filaggrin is an intermediate filament-associated protein that aggregates keratin intermediate filaments in mammalian epidermis. It is initially synthesized as a polyprotein precursor, profilaggrin (consisting of multiple filaggrin units of 324 aa each), which is localized in keratohyalin granules, and is subsequently proteolytically processed into individual functional filaggrin molecules. Active filaggrin is present at a level of the epidermis where keratinocytes are in transition between the live nucleated granular layer and the anucleate cornified layer, suggesting that filaggrin aids in the terminal differentiation process by facilitating apoptotic machinery.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.