Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filaggrin Antibody (FLG/1957R) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119377 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119377 Supplier Novus Biologicals Supplier No. NBP308651
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Filaggrin Monoclonal specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen Filaggrin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FLG/1957R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Host Species Rabbit
Immunogen Recombinant human Filaggrin protein fragment (around aa 198-288) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.