Learn More
Syngene™ FRLP Filter
GREENER_CHOICESDP
Catalog No. 01257204 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Wavelength:
670-780nm
Catalog No. Wavelength
01-257-204 670-780nm
1 options
Catalog No. 01-257-204 Supplier Syngene™ Supplier No. FILTFRLPGX
Only null left
Add to Cart
Add to Cart
Syngene™ FRLP Filter works in conjuction with the Syngene Gbox and Gbox Mini Systems and can only be used with EPI/RGB module.
  • For use in GBOX and Gbox Mini systems 7 position filter wheel.
Specifications
Wavelength 670-780nm
Intensity Fixed
For Use With (Equipment) Gbox and Gbox Mini
WARNING: Cancer - www.P65Warnings.ca.gov