Learn More
Syngene™ IR Filter
Click to view available options
Wavelength:
782-900nm
Description
- For use in GBOX and Gbox Mini systems 7 position filter wheel.
Specifications
Specifications
For Use With (Equipment) | Gbox and Gbox Mini |
Intensity | Fixed |
Wavelength | 782-900nm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov