Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GVS Filtration Membrane Gridded Nitrocellulose
Shop All Osmonics Products
Click to view available options
Pore Size:
0.45 μm
Specifications
Specifications
Autoclavable | Autoclavable |
Color | Black |
Material | MCE |
Pore Size | 0.45 μm |
Sterility | Sterile |
For Use With (Application) | Aqueous filtration; Sterility testing; Gravimetric analysis with ashing technique; Microbiological and particulate analysis; Black for food and beverage applications |
Grid Color | White |
Individually wrapped | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction