Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FIPI
GREENER_CHOICE

Catalog No. p-5545792
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Phospholipase D inhibitor

FIPI is a potent phosholipase D (PLD) inhibitor (IC50 values are 20 and 25 nM for PLD2 and PLD1 respectively). Ameliorates PLD2-driven suppression of membrane ruffling and cell spreading. Orally available.

Specifications

CAS 1781834-93-2
Quantity 10 mg
Product Type FIPI
Purity >98%
Molecular Weight (g/mol) 457.93
Inhibitors FIPI

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.