Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ FIPI

Click to view available options
Quantity:
10 mg
50 mg
Description
FIPI is a potent phosholipase D (PLD) inhibitor (IC50 values are 20 and 25 nM for PLD2 and PLD1 respectively). Ameliorates PLD2-driven suppression of membrane ruffling and cell spreading. Orally available.
Specifications
Specifications
CAS | 1781834-93-2 |
Quantity | 10 mg |
Product Type | FIPI |
Purity | >98% |
Molecular Weight (g/mol) | 457.93 |
Inhibitors | FIPI |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction