Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106271
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5106271 100 μL
1 options

Catalog No. PIPA5106271

Supplier: Invitrogen™ PA5106271

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FIS1.

Fis1, a 17KDa integral membrane protein, is a novel member of mitochondrial fission machinery. It consists of a central leucine-zipper domain, a coiled-coil region, a tetratricopeptide repeat domain and a C-terminal transmembrane domain. It is a human orthologue of Fis1p (S.cerevisiae) and is localized in the outer mitochondrial membrane. Fis1 acts as a pro-apoptotic factor and plays a vital role in the regulation of mitochondrial fission. It interacts with DLP1 and Mdv1p and facilitates Dnm1p recruitment to the functional fission complex, thereby inducing apoptosis. Role of Fis1 has also been implicated in growth and division of peroxisomes. Loss of Fis1 inhibits apoptosis and results in elongation of mammalian mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FIS1
Gene Accession No. P84817, Q9CQ92, Q9Y3D6
Gene Alias 2010003O14Rik; CGI-135; Fis1; fis1 {ECO:0000250; FIS1 homolog; fission 1 (mitochondrial outer membrane) homolog; fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae); fission 1 (mitochondrial outer membrane) homolog (yeast); fission, mitochondrial 1; H_NH0132A01.6; hFis1; Mitochondrial fission 1 protein; mitochondrial fission molecule; rFis1; tetratricopeptide repeat domain 11; tetratricopeptide repeat protein 11; TPR repeat protein 11; TTC11; UniProtKB:Q9Y3D6}
Gene Symbols FIS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FIS1(Accession Q9Y3D6), corresponding to amino acid residues Y82-M132.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288584, 51024, 66437
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.