Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Microbiology Media Additives: Tryptone

Catalog No. p-4907859 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
2000 g

Catalog No. BP1421-100

Only null left
Add to Cart

Additive for preparing bacterial nutrient media

Note: This is a product of animal origin (pancreas).
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated Powder
Grade Molecular genetics
Media Type Tryptone
Product Type Microbiology Media
For Use With (Application) Pancreatic digest of casein is used as a nitrogen source for bacteria
Packaging Type Poly Bottle
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.