Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder)
Shop All Fisherbrand Products

Click to view available options
Quantity:
500 g
2000 g
Description
- Directions for Media Preparation: Suspend 40g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Bacterial Culture |
| Form | Powder |
| Grade | Molecular genetics |
| Media Type | LB Agar |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L |
| For Use With (Application) | For preparing bacterial nutrient media |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction