Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Modified Terrific Broth (Capsules)
Shop All Fisherbrand Products
Click to view available options
Quantity:
600 g
5 kg
Description
- Directions for Media Preparation: Add 2 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Then, add 4mL of glycerol to the mixture. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Greater peptone and yeast concentration allow for higher plasmid yield
- Product is in capsule form
- Each capsule contains 24g of broth
- Format: Capsule
Specifications
Specifications
| Format | Capsule |
| Media Type | Terrific Broth |
| Product Type | Microbiology Media |
| Formulation | Casein Peptone 12g/L, Yeast extract 24g/L, Potassium Phosphate, Monobasic 2.2g/L, Potassium Phosphate, Dibasic 9.4g/L |
| For Use With (Application) | For maintenance and propagation of Escherichia coli in molecular biology procedures |
| Packaging Type | Poly pail, 24 g Capsules |
| Quantity | 5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction