Learn More
Fisherbrand™ Power Supply Adapter Set
SCI_ED
Catalog No. S67609Q Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
FisherBiotech Power Supplies
Catalog No. For Use With (Equipment)
S67609Q FisherBiotech Power Supplies
1 options
This item is not returnable. View return policy
Catalog No. S67609Q
This item is not returnable. View return policy
  • Permits electrophoresis systems with short or 90° pins to connect to the jacks of FisherBiotech Power Supplies
  • Includes one red and one black adapter
Warranty and Services
12 months
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FisherBiotech Power Supplies
Type Power Supply Adapter Set
Includes One red, one black adapter
Warranty 1 Year