Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Transfer Pipettes
GREENER_CHOICE

Catalog No. p-163759 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
4.6 mL
5 mL
Bulb Draw (Metric):
2.0 mL
3.3 mL
Length (Metric):
15 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Bulb Draw (Metric) Length (Metric)
13-711-6M 4.6 mL 3.3 mL 15 cm
13-711-5AM 5 mL 2.0 mL 15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-711-6M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Single squeeze draws up to 2.4 mL

Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from see-through low-density polyethylene.

  • Capacity: approximately 5 mL
  • Delivers 24 to 26 drops/mL
  • Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • One squeeze draws from 2.0 to 3.3 mL (depending on size used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
TRUSTED_SUSTAINABILITY

Specifications

Bulb Draw (Metric) 3.3 mL
Material LDPE
For Use With (Application) Research
Graduated Yes
Quantity 500/Pk.
Tip Style Capillary
Volume (Metric) 4.6 mL
Length (Metric) 15 cm
Type Transfer Pipet
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.