Fisherbrand™ Cell Strainers

Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

 View more versions of this product

Catalog No. 22-363-547

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Disposable Yes
    Product Type Cell Strainer
    For Use With (Equipment) All 50mL conical tubes
    Material Polypropylene
    Mesh Material Nylon
    Mesh Size 40μm
    Color Blue
    Sterile Yes

    Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

    Rapidly isolate primary cells

    • Fast, simple alternative to gauze filtration
    • Reliably dissociates cells from clumps or primary tissues
    • Consistently obtain a uniform single-cell suspension
    • Ideal for preparation of:
      • Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
      • Specimens for primary cell cultures and immunogens
      • Freezing stocks
      • Filtering agglutinative proteins produced in inactivation serum
    Reliable and easy to use
    • Strong nylon mesh with evenly spaced pores for consistent results
    • Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
    • Includes an extended lip for easier handling with forceps
    • Color-coded polypropylene frame with tab enables easy handling and identification
    • Individually packed, ready to use and disposable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit