Learn More
Fisherbrand™ Freeze Cube XL
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Purple
Includes:
30-well Cooling Blocks
96-well Cooling Blocks
Description
This cooling box is ideal for low temperature protection and collection of biological samples, can also be used to preserve unstable nucleic acid samples while working on the bench.
Specifications
Specifications
| Color | Blue |
| Includes | 30-well Cooling Blocks |
| Dimensions (L x W x H) | 152 x 170 x 123 mm |