Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Cryovial™ Storage Boxes
Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Green
Red
Yellow
Holds:
25 tubes
81 tubes
Quantity:
1 Each
4/Pk.
8/Pk.
Description
- Withstand temperatures from -196° to +121°C (-321° to +250°F)
- On 25- and 81-place boxes, a colored plastic grid insert separates the cover from the base
- The 100-place boxes feature a colored base instead of a grid insert
- Transparent cover allows view of box contents and is keyed to the base to prevent misalignment
- All boxes are supplied with a vial-removal tool to facilitate removal of vials from a full box
- Autoclavable
Specifications
Specifications
Color | Yellow |
Holds | 25 tubes |
Quantity | 1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction