Learn More
Fisherbrand™ Disposable Graduated Transfer Pipettes
Shop All Fisherbrand ProductsDescription
Graduation marks allow multiple allocations in a single draw — useful anywhere there is a need for quick, safe transfer of fluids.
Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from unbreakable, see-through low-density polyethylene to eliminate the hazards of broken glass and exposure to infectious materials.
- 5.8 mL capacity with 0.25 mL graduations
- Inert to biological fluids and most acids
- All deliver repeatable drop sizes
- Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
- Nonsterile, bulk pack of 500
Order Info
Bulk pack of 500.
Specifications
Specifications
| Bulb Draw (Metric) | 3.4 mL |
| Material | Polyethylene |
| Sterility | Non-sterile |
| For Use With (Application) | Research |
| Graduated | Yes |
| Graduations | 0.25 mL |
| Quantity | 500/Pk. |
| Packaging Type | Box of 500 |
| Volume (Metric) | 5.8 mL |
| Length (Metric) | 15.4 cm |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.