Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ General-Purpose Mercury Laboratory Thermometers: Partial-Immersion

Catalog No. p-201718
Click to view available options
Temperature Range (Metric):
-20°C to +110°C
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Available in a variety of types for a wide range of applications

  • With permanently fused markings
  • Non-roll fittings
  • Partial immersion

Order Info

Each thermometer is packaged in clear, lockable tubing.

Compliance

Made in accordance with, and meet accuracy requirements of, ANSI™/SAMA Z236.1-1983

Specifications

Length (English) Overall 12 in.
Length (Metric) Overall 305 mm
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -20°C
Temperature Range (Metric) -20°C to +110°C
Subdivision 1°C
Immersion Length 76 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.