Learn More
Fisherbrand™ Low- and Tall-Form PTFE Evaporating Dishes
Inert and usable to 280°C (536°F)
 View more versions of this product
Catalog No. 02-617-144
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Evaporating Dish
250mL
3.81 in.
Evaporating applications
50mm
100.5mm
500°F
8.45 oz.
8.45 oz., 250mL
97mm
1.96 in.
3.957 in.
PTFE
260°C