Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Polypropylene Microtiter Tubes
GREENER_CHOICE

Catalog No. p-195117 Shop All Fisherbrand Products
Click to view available options
Sterility:
Non-sterile
Sterile
Label Type:
Frosted Writing Area
No Writing Area
Packaging:
Bag
Robotic Pack
Tray Pack

Ideal for sample dilution and/or mixing prior to transfer into microtiter plates.

Ideal for sample dilution and/or mixing prior to transfer into microtiter plates

Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.

 

  • Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
  • Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
  • Frosted tubes feature writing area that wraps around tube collar
  • Capacity- 1.2mL

 

Order Info

Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks. Compact Tray PacksSnug, smooth- fitting cover protects samples and enables stacking; With paper grid for easy indentification of contentsRobotic RacksSpecifically designed with 96-well footprint for robotic workstations; One-way lid design with alphanumeric grid enables easy sample identification; Condensation rings inside cover minimize cross-contamination; Tubes fit in straight, upright position

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, ATP, DNA, pyrogen-free
Autoclavable Autoclavable
Capacity (Metric) 1.2 mL
Color Natural
Material Polypropylene
For Use With (Application) HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent
Label Type No Writing Area
Packaging Robotic Pack
Sterility Sterile
Graduated No
Product Type Microtiter Tubes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.