Learn More
Fisherbrand™ Porcelain Flat Capsule Crucibles
For use in laboratories of educational institutions, scientific development and industrial research, chemical, fertilizer, petro-chemical establishments
 View more versions of this product
Catalog No. FB960O
Add to cart
Description
Description
  • Chemical-porcelainware
  • Straight sides
  • Flatbottom
  • Glazed inside and out except for rim
  • Color: white
Specifications
Specifications
Flat Capsule Crucible
Porcelain
White
Glazed
10mm (±5.0%)
2.2mm (±0.5)
12mL
45mm (±5.0%)
Used in laboratories of educational institutions, scientific development and industrial research, chemical, fertilizer, petro-chemical establishments