Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Printed Disposable Culture Tube Caps
Shop All Fisherbrand Products
Click to view available options
Quantity:
2000/Cs.
Description
- Caps snap tight to prevent sample loss
- Blue polyethylene
- Sterile
- 500/bag (2,000/case)
Specifications
Specifications
| Product Type | Snap Cap |
| Material | Polyethylene |
| Color | Blue |
| Quantity | 2000/Cs. |
| For Use With (Equipment) | Printed Disposable Culture Tubes, 17 x 100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction