Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB10062570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062570 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062570 Supplier Novus Biologicals Supplier No. NB10062570
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

FITC Polyclonal specifically detects FITC in Non-species specific samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen FITC
Applications ELISA, Flow Cytometry
Classification Polyclonal
Conjugate HRP
Dilution Flow Cytometry 0.1 ug/mL, ELISA 1:2000 - 1:4000
Formulation 50% Glycerol/50% PBS (pH 7.4) with No Preservative
Gene Alias Fluorescein Isothiocyanate
Host Species Sheep
Immunogen Fluorescein isothiocyanate (isomer 1)
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Fluorescein isothiocyanate. This preparation has been cross-absorbed against mouse, human and rat serum proteins.
Target Species Non-species specific
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.