Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Antibody (SPM395), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB386563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386563 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386563 Supplier Novus Biologicals Supplier No. NBP308492JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FITC Monoclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ Hybridization

Specifications

Antigen FITC
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM395
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Fluorescein Isothiocyanate
Host Species Mouse
Immunogen Fluorescein isothiocyanate (FITC) conjugated to KLH
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Apoptosis Detections
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.