Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Mouse Anti-Human CD65, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569474
Description
CD65 Monoclonal antibody specifically detects CD65 in Human samples. It is validated for Flow cytometry
Specifications
CD65 | |
Monoclonal | |
FITC | |
Fucoganglioside Type II: Ceramide dodecasacharide 4c | |
Human THP-1 Cell Line | |
100 Tests | |
Human | |
Liquid |
Flow Cytometry | |
VIM8 (also known as VIM-8) | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction