Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Mouse Anti-Human CD65, BD Pharmingen™
Mouse Monoclonal Antibody
$144.00 - $372.00
Specifications
Antigen | CD65 |
---|---|
Clone | VIM8 (also known as VIM-8) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Description
CD65 Monoclonal antibody specifically detects CD65 in Human samples. It is validated for Flow cytometrySpecifications
CD65 | |
Flow Cytometry | |
FITC | |
Mouse | |
Human | |
Fucoganglioside Type II: Ceramide dodecasacharide 4c | |
IgM κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
VIM8 (also known as VIM-8) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human THP-1 Cell Line | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title