Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Rabbit, Clone: 4L8R5, Novus Biologicals™
SDP

Catalog No. NB167256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167256 20 μg
NB167255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167256 Supplier Novus Biologicals Supplier No. NBP31608020UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FITC Monoclonal antibody specifically detects FITC in All species samples. It is validated for Western Blot

Specifications

Antigen FITC
Applications Western Blot
Classification Monoclonal
Clone 4L8R5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Fluorescein Isothiocyanate
Host Species Rabbit
Immunogen FITC
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Apoptosis Detections
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.