Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FITC-Val-Ala-Asp(OMe)-FMK

FITC-Val-Ala-Asp(OMe)-FMK, Yellow Solid

$2622.00

Specifications

Concentration Peptide content: 77.6
Color Yellow
Purity >95% by TLC
For Use With (Application) Cell Viability Analysis
Content And Storage Keep cool and dry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP03FK03205
View Documents
MP Biomedicals™
03FK03205
5 mg
Each for $2,622.00
Only null left
Add to Cart
 
MP03FK03203
View Documents
MP Biomedicals™
03FK03203
3 mg N/A N/A N/A
MP03FK03201
View Documents
MP Biomedicals™
03FK03201
1 mg N/A N/A N/A
Description

Description

FITC-Val-Ala-Asp(OMe)-FMK is an inhibitor for General Caspases.
Specifications

Specifications

Peptide content: 77.6
>95% by TLC
Keep cool and dry
722.7
Yellow
Cell Viability Analysis
Proteins, Enzymes & Peptides
Solid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.