Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FITM
GREENER_CHOICE

Catalog No. p-7166821
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective negative allosteric modulator of mGlu1 receptors

FITM is a potent and selective negative allosteric modulator of mGlu1 receptors (IC50 = 5.1 nM). Displays >1300-fold selectivity for mGlu1 over other metabotropic glutamate receptors (IC50 values are 7 μM and >10 μM for mGlu5 and mGlu2 or mGlu8, respectively). Antagonizes methamphetamine-induced locomotion in mice.

Specifications

Quantity 10 mg
Molecular Formula C18H18FN5OS
Formula Weight 371.43
Product Type Negative Allosteric Modulator
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.