Learn More
Restek™ Fittings, Parker (Treated): Unions
SDP
Catalog No. 06711991 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Reducing Union
Union
Material:
Sulfinert-coated
Description:
1/16 in.
1/8 in. to 1/4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material Description
06-711-991 Union Sulfinert-coated 1/16 in.
06-711-996 Reducing Union Sulfinert-coated 1/8 in. to 1/4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-991 Supplier Restek™ Supplier No. 22520
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Type Union
Description 1/16 in.
Material Sulfinert-coated
For Use With (Equipment) Chromatography Connectors