Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIV Envelope Antibody (CE4-13B1 (13B1)), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N10663A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10663A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10663A30 Supplier Novus Biologicals Supplier No. NB10066553AF350

Mouse Monoclonal Antibody

FIV Envelope Monoclonal specifically detects FIV Envelope in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FIV Envelope
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CE4-13B1 (13B1)
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Purified gp95 envelope antigen.
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-66553 is specific for FIV Envelope gp95 protein. This antibody is directed against an epitope within the variable V2 region and cross-reacts with Petaluma, 34TF10, Ppr, MCH5-5 and Glasgow 8 isolates. NB100-66553 does not neutralize FIV and does not recognize Feline Leukaemia Virus, Feline Herpes Virus Type 1, Feline Coronavirus and Feline Calicivirus.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.