Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIV Envelope Antibody (CE4-13B1 (13B1)), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N10653PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10653PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10653PE594 Supplier Novus Biologicals Supplier No. NB10066553PEATT594

Mouse Monoclonal Antibody

FIV Envelope Monoclonal specifically detects FIV Envelope in Virus samples. It is validated for Flow Cytometry.

Specifications

Antigen FIV Envelope
Applications Flow Cytometry
Classification Monoclonal
Clone CE4-13B1 (13B1)
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Purified gp95 envelope antigen.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-66553 is specific for FIV Envelope gp95 protein. This antibody is directed against an epitope within the variable V2 region and cross-reacts with Petaluma, 34TF10, Ppr, MCH5-5 and Glasgow 8 isolates. NB100-66553 does not neutralize FIV and does not recognize Feline Leukaemia Virus, Feline Herpes Virus Type 1, Feline Coronavirus and Feline Calicivirus.
Target Species Virus
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.