Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIV gp95 Antibody (SU1-30), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB10064378Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064378Y 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064378Y Supplier Novus Biologicals Supplier No. NB10064378AF647

Mouse Monoclonal Antibody

FIV gp95 Monoclonal specifically detects FIV gp95 in Virus samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen FIV gp95
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SU1-30
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Made to FIV gp95
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64378 specifically recognizes the relatively conserved E60 peptide (amino acid sequence SSWKQRNRWEWRPDF) located within the V3 region of FIV (Feline Immunodeficiency Virus) envelope glycoprotein SU,gp95. The primary receptor for FIV infection of cats has been identified as the T-cell activation antigen CD134 which, along with CXC chemokine receptor 4 (CXCR4), facilitates the infection of CD4+ primary target cells, resulting in a depletion in the CD4+ population. Studies have shown that the V3 loop of SU,gp95 is a major immunogenic domain, suggested to contain the main determinants for the binding of FIV SU,gp95 to CXCR4.(3) Clone SU1-30 has been shown to inhibit the sCD134-activated entry of both clade B and clade C FIV strains.(1) NB100-64378 does not recognize Feline Leukaemia Virus, Feline Herpes Virus type 1, Feline Coronavirus or Feline Calicivirus. Removal of Sodium Azide is recommended prior to use in functional studies.
Reconstitution Protein A purified
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.