Learn More
Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 506
Description
- Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
- Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
- Fixable Viability Dye eFluor™ 506 can be excited by the violet (405 nm) laser line and has a peak emission of 506 nm that can be detected using a 510/50 band pass filter (equivalent to AmCyan)
- For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
- If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
- After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
- Fixable Viability Dye eFluor™ 506 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
- It should be protected from light and moisture
- Store at less than or equal to -70°C with dessicant, it may be freeze-thawed up to 20 times
- Allow vial to equilibrate to room temperature before opening
Specifications
Specifications
| Color | Cyan |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Dye |
| Emission | 405/506 |
| Product Line | eBioscience |
| Quantity | 500 Tests |
Frequently Asked Questions (FAQs)
Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.