Learn More
Invitrogen™ Fixation Medium (Medium A)
Description
The FIX & PERM™ Cell Permeabilization Kit is:
View a selection guide for all available FIX & PERM™ products and products related to sample preparation (including cell lysis and preservation solutions, blocking reagents, and information about the use of Dynabeads™ cell separation products for cell isolation).
The Fixation Medium formula was specifically designed to reduce background staining and allow simultaneous addition of fluorochrome-labeled antibodies during the permeabilization step.
How It Works
Cells are incubated for 15 minutes at room temperature with the Fixation Medium and washed. Cells can then be incubated with Permeabilization Medium (Medium B) (Cat. No. GAS002S100) and fluorochrome-labeled antibody. Both Fixation Medium and Permeabilization Medium are included in the FIX & PERM™ Cell Permeabilization Kit.
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Quantity | 5 mL |
| Volume (Metric) | 5 mL |
| Reagent Type | Fixation Reagent |
| Shipping Condition | Room Temperature |
| Product Type | Fixation Medium |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.