Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598688
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598688 Supplier Invitrogen™ Supplier No. PA598688
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FIZ1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

FLJ14768 is homologous to mouse and bovine Fiz1 (flt-3 interacting zinc finger). It interacts with NRL (neural retina leucine zipper). Possible role for Fiz1 as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIZ1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FIZ1
Gene Accession No. Q96SL8
Gene Alias AI790204; FIZ1; FLT3 interacting zinc finger 1; Flt3 interacting zinc finger protein 1; FLT3-interacting zinc finger 1; flt3-interacting zinc finger protein 1; mFLJ00416; RGD1306359; Zinc finger protein 798; ZNF798
Gene Symbols FIZ1
Host Species Rabbit
Immunogen Recombinant Human Flt3-interacting zinc finger protein 1 protein (252-410AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84922
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.