Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIZZ1/RELM alpha Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019416 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019416 Supplier Novus Biologicals Supplier No. NBP229355SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

FIZZ1/RELM alpha Polyclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen FIZZ1/RELM alpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500 -1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS
Gene Accession No. Q9EP95
Gene Alias 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2
Host Species Rabbit
Immunogen A synthetic peptide made to amino acids sequence within the residues 10-50 of the mouse RELM alpha protein.
Purification Method Immunogen affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57262
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.