Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIZZ1/RELM alpha Antibody (RM0313-9A39), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB360878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360878 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360878 Supplier Novus Biologicals Supplier No. NBP212440MFV610

Rat Monoclonal Antibody

FIZZ1/RELM alpha Monoclonal antibody specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FIZZ1/RELM alpha
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0313-9A39
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2
Host Species Rat
Immunogen Mouse recombinant FIZZ1/RELM alpha
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57262
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.