Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FK 888
SDP

Catalog No. 240010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 24-001-0 Supplier Tocris Bioscience™ Supplier No. 2400/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High affinity NK1 antagonist

Selective, high affinity tachykinin NK1 receptor antagonist (Ki = 0.69 nM) that displays 320-fold selectivity for human over rat NK1 receptors. Inhibits substance P-induced contraction of isolated guinea pig trachea (IC50 = 32 nM) and inhibits substance P-induced airway constriction in vivo following i.v. and oral administration.

Chemical Identifiers

CAS 138449-07-7
Molecular Formula C36H36N4O4

Specifications

Quantity 10 mg
Formula Weight 588.69
Purity >98%
Molecular Weight (g/mol) 588.708
Chemical Name or Material FK 888
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.