Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP10 Antibody, Novus Biologicals™
SDP

Catalog No. NB395786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395786 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395786 Supplier Novus Biologicals Supplier No. NBP24924225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP10 Polyclonal antibody specifically detects FKBP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FKBP10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias 65 kDa FK506-binding protein, 65 kDa FKBP, EC 5.2.1.8, FK506 binding protein 10 (65 kDa), FK506 binding protein 10, 65 kDa, FK506-binding protein 10, FKBP-10, FKBP6, FKBP-65, FKBP65PPIase FKBP10, FLJ20683, FLJ22041, FLJ23833, hFKBP65, Immunophilin FKBP65, OI6, peptidyl-prolyl cis-trans isomerase FKBP10, PPIASE, Rotamase
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GLPTGYLFVWHKDPPANLFEDMDLNKDGEVPPEEFSTFIKAQVSEGKGRLMPGQDPEKTIGDMFQNQDRNQDGKITV
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60681
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.