Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP14 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB086791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086791 Supplier Novus Biologicals Supplier No. NBP300140JF646

Rabbit Polyclonal Antibody

FKBP14 Polyclonal specifically detects FKBP14 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FKBP14
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM Sodium Borate
Gene Alias 22 kDa FK506-binding protein, EC 5.2.1.8, FK506 binding protein 14 (22 kDa), FK506 binding protein 14, 22 kDa, FK506-binding protein 14, FKBP-14, FKBP-22, FKBP22PPIase FKBP14, FLJ20731, peptidyl-prolyl cis-trans isomerase FKBP14,22 kDa FKBP, rotamase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FKBP14 (Q9NWM8; Met1-Lys207).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 55033
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.