Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | FKBP14 |
|---|---|
| Concentration | 0.41 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0552
![]()
|
Proteintech Group Inc
158841AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PPIases accelerate the folding of proteins during protein synthesis.Specifications
| FKBP14 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P59024, Q9NWM8 | |
| 231997, 362366, 55033 | |
| FKBP14 Fusion Protein Ag8668 | |
| Primary | |
| -20°C | |
| FKBP14 |
| 0.41 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 22 kDa FK506 binding protein, 22 kDa FKBP, FK506 binding protein 14, FKBP 14, FKBP 22, FKBP14, FKBP22, PPIase FKBP14, Rotamase | |
| Fkbp14 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title