Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP15 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292004 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292004 Supplier Novus Biologicals Supplier No. NB100424PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP15 Polyclonal specifically detects FKBP15 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen FKBP15
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate PE
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias FK506 binding protein 15, 133kDa, FK506-binding protein 133kDa, FK506-binding protein 15, FKBP133, FKBP-133, FKBP-15,133 kDa FKBP, PPP1R76,133 kDa FK506-binding protein, regulatory subunit 76, WAFL
Host Species Rabbit
Immunogen A synthetic peptide mapping to a region between residues 1175 and 1234 (C-terminus) of human FK506-binding protein 135 using the numbering given in TrEMBL entry Q9Y4D0 (GeneID 23307).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23307
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.