Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP25 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB090730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090730 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090730 Supplier Novus Biologicals Supplier No. NBP300022AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP25 Polyclonal specifically detects FKBP25 in Human samples. It is validated for ELISA.

Specifications

Antigen FKBP25
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FKBP25 (Q00688; Ala2-Asp224).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2287
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.