Learn More
Description
Specifications
Specifications
| Antigen | FKBP38 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 2.3 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:25000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | NP_036313 |
| Gene Alias | 38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase |
| Gene Symbols | FKBP8 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
