Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP51/FKBP5 Antibody (Hi51B), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB11096873U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096873U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096873U Supplier Novus Biologicals Supplier No. NB11096873UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FKBP51/FKBP5 Monoclonal specifically detects FKBP51/FKBP5 in Human, Mouse, Rat, Canine, Hamster, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FKBP51/FKBP5
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Hi51B
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q13451
Gene Alias 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, rotamase, T-cell FK506-binding protein
Gene Symbols FKBP5
Host Species Mouse
Immunogen Synthetic peptide corresponding to the residues of human FKBP51
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2289
Test Specificity Detects approx 51kDa.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.