Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP52/FKBP4 Antibody (KN382/EC1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP1974650.025MG
Description
FKBP52/FKBP4 Monoclonal specifically detects FKBP52/FKBP4 in Human, Mouse, Bovine, Guinea Pig, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
FKBP52/FKBP4 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
FKBP4 | |
Partially purified 9S rabbit uterine progesterone receptor. | |
0.025 mg | |
Membrane Trafficking and Chaperones, Stem Cell Markers | |
2288 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunohistochemistry | |
KN382/EC1 | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Guinea Pig, Primate, Human, Mouse, Hamster, Bovine, Rabbit, Mustelid | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction