Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP52/FKBP4 Antibody (KN382/EC1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

$243.50 - $731.50

Specifications

Antigen FKBP52/FKBP4
Clone KN382/EC1
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018771
SDP
View Documents
Novus Biologicals
NBP1974650.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP19746505
SDP
View Documents
Novus Biologicals
NBP1974650.05MG
0.05 mg
Each for $366.00
Only null left
Add to Cart
 
NBP19746502
SDP
View Documents
Novus Biologicals
NBP1974650.2MG
0.2 mg
Each for $731.50
Only null left
Add to Cart
 
Description

Description

FKBP52/FKBP4 Monoclonal specifically detects FKBP52/FKBP4 in Human, Mouse, Bovine, Guinea Pig, Hamster, Mustelid, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

FKBP52/FKBP4
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Monoclonal
Purified
RUO
Guinea Pig, Primate, Human, Mouse, Hamster, Bovine, Rabbit, Mustelid
51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase
FKBP4
IgG1
Protein G purified
KN382/EC1
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunohistochemistry
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Stem Cell Markers
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
2288
Partially purified 9S rabbit uterine progesterone receptor.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.