Learn More
Description
Specifications
Specifications
| Antigen | FKBP6 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 9B7 |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase |
| Gene Symbols | FKBP6 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
